missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets

Catalog No. 75008133
Change view
Click to view available options
Color:
Green
Light Gray
Olive Brown
Red
White
Yellow
Tube Size:
10/15 mL
175/250 mL
7 mL
15 mL
50 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Tube Size
75008133 Yellow 7 mL
75008134 Light Gray 7 mL
75008135 Red 15 mL
75008136 White 10/15 mL
75008137 Olive Brown 15 mL
75008140 Green 50 mL
75008144 White 175/250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 75008133 Supplier Thermo Scientific™ Supplier No. 75008133

For use with four-place rotors 75006445 and 75006434 when equipped with round bucket 75006441

Adapters sold individually; order correct quantity to equip your rotor.

Specifications

For Use With (Equipment) LH-4000W and LH-4000 rotor with round bucket
Color Yellow
Tube Size 7 mL