missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Four-Place Swing Bucket Bioshield Rotor

Catalog No. 75006461
Change view
Click to view available options
Capacity:
20 x 7 mL Tubes
4 x 50 mL Conical Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75006461 4 x 50 mL Conical Tubes
75006454 20 x 7 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75006461 Supplier Thermo Scientific™ Supplier No. 75006461

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with four-place Bioshield rotor 75-006-435 and with four-place rotor 75-006-445 when equipped with rectangular bucket 75-006-446

Adapters sold individually; order correct quantity to equip rotor.

Specifications

For Use With (Equipment) LH-4000W and LH-4000 rotor with DoubleSpin bucket
Capacity 4 x 50 mL Conical Tubes
Product Type Tube Adapter
Color Light Green