Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
Supplier:  Thermo Scientific™ 75008186
 View more versions of this product
Catalog No. 75008186
Only null left
Description
Description
  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifications
Chemical Identifiers
4 x 180 Swing bucket rotor
Specifications
4 x 180 Swing bucket rotor
Red
Product Suggestions
Product Suggestions