missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor

Catalog No. 75008183
Change view
Click to view available options
Color:
Brown
Gray
Green
Green/Yellow
Red
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
75008183 Green/Yellow
75008182 Green
75008187 Brown
75008185 White
75008186 Red
75008188 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 75008183 Supplier Thermo Scientific™ Supplier No. 75008183

For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172

  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes

Order Info

Adapters are sold individually; order correct quantity to equip rotor.

Specifications

For Use With (Equipment) 4 x 180 Swing bucket rotor
Color Green/Yellow