missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor

Catalog No. 75008182
Change view
Click to view available options
Color:
Brown
Gray
Green
Red
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
75008182 Green
75008187 Brown
75008185 White
75008186 Red
75008188 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 75008182 Supplier Thermo Scientific™ Supplier No. 75008182
Only null left

For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172

  • Accommodate tube sizes from 1.5 to 100mL, including urine tubes

Order Info

Adapters are sold individually; order correct quantity to equip rotor.

Specifications

For Use With (Equipment) 4 x 180 Swing bucket rotor
Color Green