missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Mucolexx™

Catalog No. 2861211
Change view
Click to view available options
Quantity:
4 x 3.8L (1 gal.)
4 x 500mL (0.13 gal.)
50 x 120mL, filled to 60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2861211 50 x 120mL, filled to 60mL
2861210 4 x 500mL (0.13 gal.)
2861209 4 x 3.8L (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2861211 Supplier Epredia™ Supplier No. 9990375
Only null left

 

  • Ready-to-use mucoliquefying preservative
  • Liquefies (with agitation) mucoid cytology specimens
  • Maintains cellular detail
  • Short-term preservation of specimens

 

  • Histology
  • Cytology

Specifications

Color Light Red
Packaging Type Cups
Product Type Tissue Preservative
Quantity 50 x 120mL, filled to 60mL
For Use With (Application) Provides short term preservation of specimens
Form Liquid