missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Mucolexx™
Description
- Ready-to-use mucoliquefying preservative
- Liquefies (with agitation) mucoid cytology specimens
- Maintains cellular detail
- Short-term preservation of specimens
- Histology
- Cytology
Specifications
Specifications
| Color | Light Red |
| Product Type | Tissue Preservative |
| Quantity | 4 x 500mL (0.13 gal.) |
| For Use With (Application) | Provides short term preservation of specimens |
| Form | Liquid |