missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Consul-Mount™

Catalog No. 9990440
Change view
Click to view available options
Product Type:
Cytology Media
Mountant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
9990440 Mountant
9990441 Cytology Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 9990440 Supplier Epredia™ Supplier No. 9990440

Designed specifically for use with the Epredia™ Consul™ coverslipper

  • Permanent, organic mountant
  • Xylene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (125-200 cps at 24°C)

Specifications

Color Clear
Product Type Mountant
Quantity 500 mL
For Use With (Application) For use with Epredia™ Consul™ coverslipper
Form Liquid