Learn More
Thermo Scientific™ Round Buckets for 4-Place Swinging Bucket Rotor
Catalog No. 75007555
Change view
Click to view available options
For Use With (Equipment):
Swinging bucket rotor
Catalog No. For Use With (Equipment)
75007555 Swinging bucket rotor
1 options
Catalog No. 75007555 Supplier Thermo Scientific™ Supplier No. 75007555
4 x 100mL capacity buckets fit the Thermo Scientific Swinging 4-place rotor (75-007-591)
Order Info
Sold individually; order correct quantity to equip rotor. Biocontainment lids (75-007-598) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5 to 50mL. No adapter is required for 100mL bottle.
Specifications
For Use With (Equipment) Swinging bucket rotor