missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ STS™ Specimen Transport System Pre-filled Specimen Containers in Handi-pack Configuration, 10% NBF

Handi-pack packaging configurations are specimen containers in smaller, re-usable and easy to carry trays. They are ideal for satellite offices that collect specimens to be couriered to a central laboratory.

Supplier:  Epredia™ 511201

 View more versions of this product

Catalog No. 361015676


Only null left

Description

Description

Specifications

Chemical Identifiers

Pre-filled Specimen Container
Screw Top
96/Cs.
Polypropylene
10% NBF

Specifications

Pre-filled Specimen Container
120 mL, pre-filled 60 mL
10% NBF
Polypropylene
Screw Top
96/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions