missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ STS™ Specimen Transport System Pre-filled Specimen Containers in Handi-pack Configuration, 10% NBF
Handi-pack packaging configurations are specimen containers in smaller, re-usable and easy to carry trays. They are ideal for satellite offices that collect specimens to be couriered to a central laboratory.
Supplier: Epredia™ 51901
Chemical Identifiers
| Pre-filled Specimen Container | |
| Screw Top | |
| 96/Cs. |
| Polypropylene | |
| 10% NBF |
Specifications
| Pre-filled Specimen Container | |
| 90 mL, pre-filled 45 mL | |
| 10% NBF |
| Polypropylene | |
| Screw Top | |
| 96/Cs. |