missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ CTA w/1% Maltose

Catalog No. R060620
Change view
Click to view available options
Quantity:
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060620 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R060620 Supplier Thermo Scientific™ Supplier No. R060620

For determination of fermentation reactions by fastidious microorganisms, such as Neisseria, pneumococci, streptococci, and non-spore forming anaerobes.

Specifications

Additive 1% Maltose
Media Type CTA Agar (Cystine Trypticase Agar)
Product Type Microbiology Media
For Use With (Application) Determination of fermentation reactions by fastidious microorganisms such as &italic_begin;Neisseria&italic_end;, pneumococci, streptococci, and nonspore forming anaerobes
No. per Pack 20
Quantity 4 mL