Learn More
Thermo Scientific™ Remel™ Bordet Gengou Agar (with blood)
Catalog No. R01240
Change view
Click to view available options
Bordet Gengou Agar w/Blood
Catalog No.
R01240 Bordet Gengou Agar w/Blood
1 options
Catalog No. R01240 Supplier Thermo Scientific™ Supplier No. R01240
With blood; For cultivation and isolation of Bordetella pertussis and other Bordetella spp.
Specifications
Certifications/Compliance USDA, APHIS
Manufacturing Origin United States
Media Type Bordet Gengou Agar with Blood
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Packaging Type Plate
Quantity 10/Pk.