Learn More
Thermo Scientific™ Reacti-Vial Reaction Vials
An internal cone makes small sample handling easy and convenient
Supplier: Thermo Scientific™ TS13223
Description
- Extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions
- Amber vials available for light-sensitive compounds
- Supplied complete with Open-Top screw caps and PTFE/Rubber discs
Residue isolation, Derivative preparation, Maximum sample retrieval, Moisture protection, Sample storage, Precipitations, Centrifugations, Solvent evaporation
Specifications
| Clear | |
| 5 mL | |
| 5 mL | |
| Certificate of Analysis (COA) |
| Silica Glass | |
| Internal | |
| Screw Cap | |
| Vial |