missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysozyme

Catalog No. PI89833
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89833 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89833 Supplier Thermo Scientific™ Supplier No. 89833

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Degrade unwanted bacterial cell wall in protein or nucleic acid extracts

Lysozyme is an enzyme used to break down bacterial cell walls to improve protein or nucleic acid extraction efficiency. Lysozymes (muramidases) are a family of enzymes with antimicrobial activity characterized by the ability to damage the cell wall of bacteria. The enzyme acts by catalyzing the hydrolysis of 1,4-beta-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between the N-acetyl-D-glucosamine residues in chitodextrins. Although hen egg white lysozyme is most effective for the lysis of gram-positive bacteria, it also facilitates the lysis of gram-negative bacteria such as Salmonella and Shigella. The lysis of E. coli is especially improved by the addition of both lysozyme and a nucleases such as DNase I.

Highlights:

  • Bacterial cell wall lytic enzyme that improves inclusion body protein purification
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • Available in two convenient formats: lyophilized powder for large volume applications (Part No. 89833) and convenient glycerol freezer stock (Part No. 90082)

Properties of lysozyme:

  • Enzymatic function: Catalyzes the hydrolysis of 1,4-β-linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues in peptidoglycans and between N-acetyl-D-glucosamine residues in chitodextrins
  • Molecular weight: 14.388 kDa
  • Extinction coefficient: ∽26.4 at 280nm
  • Isoelectric point: pH 11.0
  • Unit definition: 1 unit is that amount of enzyme needed to catalyze a decrease in absorbance at 450nm of 0.001/min at 25 degrees at pH 6.24 in a 1cm cuvette due to lysis using a ∼0.25mg/mL suspension of Micrococcus luteus.

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Upon receipt, store at -20°C in a nonfrost-free freezer. Product shipped with ice.
Form Lyophilized
Reagent Type Enzyme for Cell Lysis
Enzyme Lysozyme (Muramidase)
Quantity 5 g
How should I prepare and store Lysozyme (Cat. No. 89833)?

Lysozyme powder can be dissolved in 0.1 M KH2PO4 (pH 6.24) to the desired concentration - up to 5 mg/mL. Once the solution has been prepared, it can be stored at 4 degrees C for up to 5-6 weeks, or at room temperature for up to 6-7 days, without affecting performance.

Can you explain how the B-PER Bacterial Protein Extraction Reagent lyses cells?

The B-PER Reagent solution contains a proprietary, mild, non-ionic detergent in 20 mM Tris-HCl, pH 7.5. It effectively disrupts cells and solubilizes native or recombinant proteins without denaturation. The reagent creates holes in the cell membrane that will leak out cytosolic proteins. The sample may become very viscous when the bacterial chromosome is released. We recommend adding DNAse I (Cat. No. 90083) to the reagent to reduce viscosity. For better lysis efficiency and if there are inclusion bodies, we recommend adding Lysozyme (Cat. No. 90082) to the reagent. Alternatively, you may purchase the B-PER Bacterial Protein Extraction Reagent with Enzymes Kit (Cat. No. 90078 or 90079) that includes the B-PER Bacterial Protein Extraction Reagent, DNase I, and Lysozyme.

For Research Use Only. Not for use in diagnostic procedures.