missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HA Synthetic Peptide
Description
HA Synthetic peptide 26184 from Thermo Scientific™ specifically detects HA in samples, and it is validated for Control
The HA peptide, NH2-YPYDVPDYA-COOH, is a synthetic peptide used to competitively elute fusion proteins containing the influenza hemagglutinin (HA) tag bound to immobilized anti-HA affinity resins.
Formulation: 5 mg purified Peptide (1mg/mL) no preservative;
26184 detects HA in samples. 26184 has been successfully used in Control procedures.
Formulation: 5 mg purified Peptide (1mg/mL) no preservative;
26184 detects HA in samples. 26184 has been successfully used in Control procedures.
Specifications
Specifications
| Protein Form | Recombinant |
| Content And Storage | Store at 4°C. |
| For Use With (Application) | Immunoprecipitation |
| Quantity | 5 mg |
Frequently Asked Questions (FAQs)
Once the HA Synthetic Peptide (Cat. No. 26184) is resuspended, at what temperature should I store it?
After resuspending the HA Synthetic Peptide at 1 mg/mL in TBS or water, we recommend aliquoting it and freezing at -20 degrees C. Avoid multiple freeze-thaws.
For Research Use Only. Not for use in diagnostic procedures.