Learn More
Fisherbrand™ accumet™ Chloride Combination Electrode - Mercury-Free
Catalog No. 13620627
Change view
Click to view available options
Connector Types:
BNC
Catalog No. Connector Types
13620627 BNC
1 options
Catalog No. 13620627 Supplier Fisherbrand™ Supplier No. 13 620 627
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Typical applications include water/wastewater, soil, dairy, meats, tomato/vegetable products.
Method ASTM/AOAC approved
Specifications
Applications Chloride
Connector Types BNC
Junction Sure-Flow Junction
Material Plastic
For Use With (Equipment) ISE Meter with a BNC Connection
Temperature (Metric) Operating 0°C to 80°C
Type Chloride Combination Electrode