Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
Catalog No. 12565364
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Lid:
With Lid
Without Lid
Skirt Style:
High Flange
Low Flange Pinchbar
Barcode:
Includes Barcode
No Barcode
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Lid Skirt Style Barcode
12565364 Sterile With Lid High Flange Includes Barcode
12565362 Non-sterile Without Lid High Flange Includes Barcode
12565210 Sterile Without Lid Low Flange Pinchbar No Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12565364 Supplier Thermo Scientific™ Supplier No. 260887
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Barcode Includes Barcode
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format Standard
Lid With Lid
Description 96F well plate, Non-Treated, clear, with lid, Sterile, Barcoded (Pack of 10, Case of 160)
Array 8 x 12
Packaging 10 Pk.
Product Line Nunc
Color Clear
Product Type Microplate
Skirt Style High Flange
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
No. per Case 160
No. per Pack 10
Quantity 160/Cs.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.