Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
Through-the-top tubing with closure and stem molded in one piece
Supplier:  Thermo Scientific™ 7124010001
 View more versions of this product
Catalog No. 02-923-17A
Only null left
Description
Description
  • Closure and stem molded in one piece for leakproof service
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Leakproof†
Specifications
Specifications
Polypropylene
28-400
Wash bottle
Product Suggestions
Product Suggestions