Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
Description
- Closure and stem molded in one piece for leakproof service
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Leakproof†
Specifications
Specifications
| Material | Polypropylene |
| Neck Finish | 24-400 |
| For Use With (Equipment) | Wash bottle |