missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Centrifuge Bottle Sealing Cap Assembly

Catalog No. 03313108
Change view
Click to view available options
Includes:
Blue Silicone Gasket
Red Silicone Gasket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
03313108 Blue Silicone Gasket
03313109 Red Silicone Gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03313108 Supplier Thermo Scientific™ Supplier No. DS3132BLUE

Includes

Includes polypropylene screw cap and shoulder ring, Tefzel™ ETFE plug, and Viton™ fluoroelastomer O-ring to form a leakproof† seal

Seals centrifuge bottle

  • For use with 1L centrifuge bottles
  • Autoclavable
  • Polypropylene screw closure
  • For use at recommended maximum speeds (10,000 x g or greater) without leakage, even when full

Specifications

Color Blue
Includes Blue Silicone Gasket
Material Polypropylene
For Use With (Equipment) 1 L PC Nalgene Centrifuge Bottle