missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Low Profile Tubes and Flat Caps, strips of 8, assorted colors

Catalog No. AB0778
Change view
Click to view available options
Quantity:
250 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0778 250 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0778 Supplier Thermo Scientific™ Supplier No. AB0778

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Low profile strip tubes for PCR and qPCR applications.

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
Applications
  • PCR
  • qPCR/Real-Time PCR

Hazardous: No
Storage Condition: Ambient

PCR, qPCR and Real-Time PCR.

Certifications

Certified free from human DNA, RNases, and DNases.

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Assorted
Volume (Metric) Working 0.1 mL
Volume (Metric) 0.15 mL
Quantity 250 strips
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Reaction Speed Low profile
Closure Type Flat Cap
Closure Included Flat Caps
Format 250 Strip Tubes and Caps
Product Type Low Profile Tube and Flat Cap
Show More Show Less
When should I use a Thermo Scientific low profile PCR plastic?

We recommend Thermo Scientific low profile PCR plates and tubes for reaction volumes below 20 µL. The low profile design minimizes the airspace between the PCR reaction and reduces evaporation effects, thereby increasing PCR efficiency in low volume reactions.

What is a low profile Thermo Scientific plate/tube?

Low profile Thermo Scientific PCR plastics have shorter wells than the standard profile plastics, decreasing the “dead space” between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction.

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.