Learn More
Thermo Scientific™ Adjustable Reinforced Shelf with Channels
Description
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
Specifications
Specifications
| Description | Reinforced Stainless Steel Shelf and Channels (max. load 68kg) |
| For Use With (Equipment) | Large Capacity Reach-In CO2 Incubator |
| Lighting | LED |
| Material | Stainless Steel |
| Type | Shelf |