Learn More
Thermo Scientific™ Adapters for HBB-6 Swinging Bucket Rotor
For use with the Thermo Scientific HBB-6 Swinging Bucket Rotor
Supplier:  Thermo Scientific™ 01094
 View more versions of this product
Catalog No. 01-094
Only null left
Explore available promotions
Description
Description
  • A range of adapters to accommodate a variety of blood bags varying in sizes.
  • Adapters for up to 12 platelet-rich plasma and buffy coat bags
Specifications
Specifications
Adapter, 450 to 550 mL
6 x 550 mL
Double or Triple Blood Bag
6
Product Suggestions
Product Suggestions