Learn More
Thermo Scientific™ HBB-6/HLR-6 Bucket Adapters
Enable blood bag centrifuging in the Sorvall RC3BP
$355.65
Description
For use in the HBB-6/HLR-6 rotors.
- Adapters accept triple, quint or quad blood bags
- Maximum capacity: 6 x 550mL