missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NTP Set, Tris buffered

Catalog No. FERR1481
Change view
Click to view available options
Quantity:
4 x 0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1481 4 x 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1481 Supplier Thermo Scientific™ Supplier No. R1481
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions.

Thermo Scientific NTP Set, Tris buffered is a convenient set of 100 mM ATP, CTP, GTP, and UTP solutions. These extremely stable nucleotides are supplied as aqueous solutions titrated to pH 7.3-7.5 with Tris base. The nucleotides have greater than 99% purity and are free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields with Tris buffered NTPs when compared to NTPs titrated with NaOH
• Stable for 3 years at -20°C, and after multiple freeze-thaw cycles

Applications
• in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

Related products
• ATP, Tris Buffered (Cat. No. R1441)
• GTP, Tris Buffered (R1461)
• UTP, Tris Buffered (R1471)

Specifications

Content And Storage NTP Set, Tris buffered, 100 mM (4 x 0.25 mL)

Store at –20°C.
Shipping Condition Dry Ice
Concentration 100 mM
Label or Dye Unlabeled
Quantity 4 x 0.25 mL
For Use With (Application) in vitro Transcription, RNA Amplification
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.