Learn More
Thermo Scientific™ Evolution™ One, OnePlus and Pro Spectrophotometer Solid Sample Holders
Designed to hold solid samples for transmittance measurements
Supplier: Thermo Scientific™ 222217200
Description
Designed to hold solid samples for transmittance measurements
Solid Sample Holder
- Provides easy mounting of solid samples like filters, plastic, glass and optics
- Accommodates standard 2 x 3 in. sample holders
- Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
- Requires one Solid Sample Holder (14-387-503)
Chemical Identifiers
| Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers |
Specifications
| Solid Sample Holder | |
| Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers | |
| Std 2 x 3 in. Sample Holders |