missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Easy Pierce Heat Sealing Foil, 38 μm
Description
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
- Suitable for PCR (requires the pressure from a screw-down heated lid)
- Easily pierced
- Peelable bond can be removed by hand
- 20μm foil is even more easily pierced and is suitable for multiple layering
- Recommended for use with DMSO
- Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)
Specifications
Specifications
| Material | Aluminum |
| Dimensions (L x W) | 80 x 125 mm |
| Quantity | 100/Pk. |
| Format | Sheets |
| For Use With (Equipment) | ALPS 25 Automated Laboratory Plate Sealer |
| Product Type | Easy Pierce Foil |
| Thickness | 38 μm |