Learn More
Thermo Scientific™ CryoMed™ Thermocouple Sensors
Sample thermocouple sensors
$309.98 - $859.42
Description
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
- Include male plug
For controlled-rate freezing applications