Learn More
Thermo Scientific™ CryoMed™ Bag Freezing Presses
Catalog No. 15467420
Change view
Click to view available options
Holds:
Fenwal 4R5461 (2 Bags)
Fenwal 4R5461 (4 Bags)
Gambro DF-200 (2 Bags)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
15467420 Fenwal 4R5461 (4 Bags)
15465693 Fenwal 4R5461 (2 Bags)
13262819 Gambro DF-200 (2 Bags)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15467420 Supplier Thermo Scientific™ Supplier No. 4000318
CryoMed™ Bag Freezing Presses
Bag presses for Nexell, Fenwal, Gambro and Delmed. Bag capacities 2-4 bags.
Specifications
Type Freezing Press
For Use With (Equipment) CryoMed
Holds Fenwal 4R5461 (4 Bags)