missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Seal Heat Sealing Film, For ALPS 300 and 3000; Roll

Catalog No. AB3797
Change view
Click to view available options
For ALPS 300 and 3000; Roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AB3797 For ALPS 300 and 3000; Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3797 Supplier Thermo Scientific™ Supplier No. AB3797

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for applications requiring pierceable non-conductive seal material

  • Suitable for PCR applications with screw- or clip-down heated lid thermal cyclers only
  • Seals polypropylene, polystyrene, polyethylene and Diamond plates
  • Easily pierced and peelable by hand from 96-well plates
  • Temperature range: -80°C to +80°C

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description Clear Seal Thin, Polymer, 78mm x 610m
Material Polyethylene
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
Length (English) Clear Seal Thin, Polymer, Roll 78mm x 610m
No. per Case 1 Roll
Dimensions (L x W) 610 m x 78 mm
For Use With (Application) Storage, PCR, qPCR, Automation, ELISA, Molecular Diagnostics
Format Roll
For Use With (Equipment) ALPS 5000 & ALPS 3000
Product Type Heat Seal
Product Line Abgene
Adhesive Polypropylene
Certified RNase/DNase-free No
Pierceable Yes
Temperature Range (Metric) -80°C to +80°C, -80°C to +120°C (in Cycler)
Well Shape All well shapes
Plate Compatibility Polypropylene, Polyethylene, Polystyrene
Show More Show Less