missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Seal Heat Sealing Film

Ideal for applications requiring pierceable non-conductive seal material

$8169.09 - $8169.09

Specifications

Color Clear
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description Clear Seal Thin, Polymer, 78mm x 610m
Material Polyethylene
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Format Price Quantity  
Catalog Number Mfr. No. Format Price Quantity  
AB-3797
View Documents
Thermo Scientific™
AB3797
Roll
Each for $8,169.09
Only null left
 
Description

Description

Ideal for applications requiring pierceable non-conductive seal material

  • Suitable for PCR applications with screw- or clip-down heated lid thermal cyclers only
  • Seals polypropylene, polystyrene, polyethylene and Diamond plates
  • Easily pierced and peelable by hand from 96-well plates
  • Temperature range: -80°C to +80°C

Specifications

Specifications

Clear
Clear Seal Thin, Polymer, 78mm x 610m
Non-sterile
96
1 Roll
Storage, PCR, qPCR, Automation, ELISA, Molecular Diagnostics
ALPS 5000 & ALPS 3000
Abgene
No
-80°C to +80°C, -80°C to +120°C (in Cycler)
Polypropylene, Polyethylene, Polystyrene
Life Science Reagent Resistant
Polyethylene
Not Autoclavable
Clear Seal Thin, Polymer, Roll 78mm x 610m
610 m x 78 mm
Roll
Heat Seal
Polypropylene
Yes
All well shapes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'