Learn More
Thermo Scientific™ Chemically-Preserved Environmental Sample Containers
Provide an extra measure of assurance that proper sampling protocols are followed
Supplier: Thermo Scientific™ PP14040CEP5ST
Includes: Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.
Description
- Standard Thermo Scientific Certified sample containers chemically-preserved with certified chemical preservatives for environmental sample analyses
- Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport
Custom preserved containers available by special order:
- Choose a standard certified Thermo Scientific environmental sample container
- Choose a chemical from our standard chemical list
- Specify the volume and concentration of chemical to aliquot into each container
- 10 Case minimum order
Order Info
Products available only in the US
Specifications
| Clear | |
| 40 mL clear septa vial w/0.5 mL sodium thiosulfate | |
| 24-414 |
| Environmental Sampling | |
| Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification for highway and rail transport |