Learn More
Thermo Scientific™ Biological Indicators
Catalog No. 11715140
Change view
Click to view available options
Quantity:
100 Vials/Box
25 Vials/Box
Population:
106 Spores
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Population
11715140 25 Vials/Box 106 Spores
11715140A 100 Vials/Box 106 Spores
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11715140 Supplier Thermo Scientific™ Supplier No. AY759X7
Only null left
Easy and inexpensive methods to monitor steam sterilizer effectiveness
Indicators verify sterility of instruments, vials, media, liquid cycles and other products in less time than it takes to send samples to an outside lab. Choice of two biological indicators: for media and liquids or for instruments, glassware, and other applications. Color change from purple to yellow indicates an ineffective sterilization process.
For Media and Liquids
  • SterilAmp™ II Biological Indicators with spore populations of 106
  • Available in boxes of 25 or 100
For Instruments, Glassware, and Other Applications
  • B/T Sure™ Biological Indicators with spore populations of 105 Other spore populations available upon request Comply with current AAMI and ISO standards
B/T Sure and SterilAmp II Biological Indicators
  • Verify sterility of instruments, media, and other steam-sterilized items in just two days, using a simple three-step process
  • Easy sterilization identification: Purple color ensures sterilization; yellow, cloudy results indicate an ineffective sterilization
  • Indicators can save up to 60% of the cost of outsourcing complete sterility tests
  • B/T Sure indicators are for instruments, glassware, and other apparatus; have spore population of 105
  • SterilAmp II indicators are for media and additives; have spore population of 106
Chemitest™ Temperature Indicators
  • Verify that sterilizer reached adequate sterilization temperature during a cycle
  • Can also be used to verify that an item has been run through a sterilization cycle
  • Available in pouches, tabs, strips, and tape
Specifications
Description Sterilamp II Biological Indicator
Type Accessory, Indicator
For Use With (Application) Media and additives
Population 106 Spores
Quantity 25 Vials/Box
Sterilization Indicator Yes