missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ ROpure LP™ Cartridges

Catalog No. 09050276
Change view
Click to view available options
Cartridge Ultrapure No Spring
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
09050276 Cartridge Ultrapure No Spring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09050276 Supplier Thermo Scientific™ Supplier No. D 5027

Thermo Scientific Barnstead; For Use with: Nanopure Quad, E-pure; Cartridge: Ultrapure Ion Exchange

Pretreatment/Prefilter, Thin Film

  • Extruded carbon cartridge (with 5μm prefilter) extends life of 09-050-37 membrane by removing chlorine from feedwater
  • Ultrapure Ion Exchange
    • Capacity: 760 grains as NaCl to 1megohm endpoint
    • Can also be used with E-pure™ systems and Nanopure™ systems

    Specifications

    Capacity 760 grains
    Purification Method Ultrapure
    For Use With (Application) Spring ion exchange
    For Use With (Equipment) Nanopure Quad and E-Pure System, Spring Ion Exchange
    Type Cartridge