Learn More
Thermo Scientific™ Barnstead Nanopure Consumables Packages
Save up to 15% off list prices on Nanopure cartridges and accessories
Supplier:  Thermo Scientific™ D502143
 View more versions of this product
Catalog No. 11500032
Only null left
Includes: Cartridge for R/O or distilled water feed (D50280), UV replacement lamp (LMX13), and sanitization kit (CMX25). Life Science packages add replacement ultrafilter membrane (FL1192X1).
Description
Description
For use with Nanopure systems. Choose from UV or Life Science packages for R/O distilled or deionized feeds.
Nanopure UV Package for R/O Distilled Feed
  • Includes Durapack™ organic-free R/O cartridge pack, UV replacement lamp and sanitization kit
  • Save 13%
Nanopure UV Package for Deionized Feed
  • Includes Durapack™ organic-free deionization cartridge pack, UV replacement lamp and sanitization kit
  • Save 12%
Nanopure Life Science Package for R/O Distilled Feed
  • Includes Durapack™ organic-free R/O cartridge pack, UV replacement lamp, sanitization kit and replacement ultrafilter membrane
  • Save 15%
Nanopure Life Science Package for Deionized Feed
  • Includes Durapack™ organic-free deionization cartridge pack, UV replacement lamp, sanitization kit and replacement ultrafilter membrane
  • Save 15%
Specifications
Specifications
Nanopure Systems w/Deionized Feed
Durapack™ organic-free deionized cartridge pack, UV replacement lamp, sanitization kit and replacement ultrafilter membrane
Nanopure Life Science Package
Product Suggestions
Product Suggestions