missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ART Filtered Pipette Tips in Racks in Lift-off Lids
Description
Pipette samples with the accuracy and precision you expect from your pipette, every time with SoftFit-L™ Filtered Pipette Tips in Racks in Lift-off Lids. For use with Rainin™ LTS pipettes.
The SoftFit-L System is compatible with Rainin LTS pipettes made by Rainin Instrument LCC.
- Available in low retention
- Sterile tips are certified free from RNase, DNase and pyrogen
- Manufactured according to the industry's highest quality standards
Specifications
Specifications
| Volume (Metric) | 500 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
| Format | Rack |
| For Use With (Equipment) | Rainin LTS LiteTouch |
| Product Line | ART |
| Fit for Purpose | Sterile & PCR Ready |