missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Polystyrene 96-Well Plate Cover with Notches
GREENER_CHOICE

Catalog No. AB0751
Change view
Click to view available options
Plate Compatibility:
Abgene 96 and 384-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plate Compatibility
AB0751 Abgene 96 and 384-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0751 Supplier Thermo Scientific™ Supplier No. AB0751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking

Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates. The covers are made of polystyrene or polypropylene and are either ridged or non-ridged.

Highlights

  • Polystyrene covers can be used with all 96-well plates
  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates

 

Hazardous: No

Storage Condition: Ambient

Specifications

Color Clear
Description Microplate Lid
Dimensions (L x W x H) 5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm)
Material Polystyrene
Sterility Cleanroom
No. of Wells 96
Includes Without stacking ridge (for automated systems)
Condensation Rings No
No. per Case 50
No. per Pack 2
Dimensions (L x W) 5 x 3.31 in. (126 x 84 mm)
Format Lid
Product Type Microplate Lid, cover
Product Line Abgene
Adhesive None
Temperature Range (Metric) -20°C to +120°C
Well Shape All well shapes
Plate Compatibility Abgene 96 and 384-well Plate
Cut-off Corners H1 Corner
Fit for Purpose PCR Ready
Show More Show Less