missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 48 Well 6 mL Polypropylene Storage Plate
GREENER_CHOICE

Catalog No. AB0988
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
AB0988 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0988 Supplier Thermo Scientific™ Supplier No. AB0988

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The 6 mL 48-well storage plate is ideal for storage when large sample volumes are required.

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

Abgene 48 Well 6 mL Polypropylene Storage Plate are manufactured in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free.

Highlights

  • 6 mL working volume when sealed with adhesive or heat seals
  • 48 square wells
  • Pyramidal well bottom for ease of sample retrieval
  • Support and stacking ribs
  • Angled well corners reduce the capillary action of liquids, preventing cross-contamination between wells

Applications

  • Sample storage
  • Cell culture
  • Assay setup; diluting and aliquoting

Compatible Sealing Film:Adhesive and Heat

Hazardous: No

Max Centrifugal Force: Not suitable for centrifugation

Max Vol (Cap Seal): n/a

Max Vol (Film Seal): 6 mL

Max Vol (Mat Seal): n/a

Storage Condition: Ambient

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 48-well Polypropylene Sample Processing & Storage Plates
Array 6 x 8
Autoclavable Autoclavable
Packaging Individually Wrapped
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 48-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 6000 μL
No. of Wells 48
Volume (Metric) Working 5500 μL
Sterility Sterile
Surface Treatment Untreated
Well Shape Square, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case 50
No. per Pack 1
Regulatory Status RUO
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.