missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film

Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 200 L |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 55.9 cm x 91.4 cm (22 in. x 36 in.) |
| Quantity | 20 per case |
| For Use With (Equipment) | SV30113 |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | HyPerforma |
| Port Detail | 0 Ports |
| Show More |