missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, strips of 8,white

Catalog No. AB0264W
Change view
Click to view available options
Quantity:
250 strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0264W 250 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0264W Supplier Thermo Scientific™ Supplier No. AB0264W

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color White
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 250 strips
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Reaction Speed Standard
Closure Type Not Included
Format 250 Strip Tubes
Product Type 8-strip Tube
Show More Show Less