missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, 0.2 mL, domed cap

Catalog No. AB0337
Change view
Click to view available options
Quantity:
1000 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0337 1000 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0337 Supplier Thermo Scientific™ Supplier No. AB0337

0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors

0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.

The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed

Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage

Hazardous:No
>Storage Condition:Ambient

PCR; Sample storage

Certifications

Certified free from human DNA, RNases, and DNases

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Natural
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 1000 tubes
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Reaction Speed Standard
Closure Type Domed Cap
Format 1,000 Tubes
Product Type Tube
Show More Show Less