This filter set is designed for the CFP-YFP FRET assay, providing specific excitation and emission wavelengths for accurate fluorescence resonance energy transfer measurements.
- Excitation wavelength/FWHM: 438/30 nm
- Emission wavelength/FWHM: 497/21 nm
- Acceptor emission wavelength/FWHM: 542/25 nm
- Optimized for CFP-YFP FRET applications