Learn More
Corning™ ThermalTray™ Thermo-conductive Platforms
Support Corning CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths, and liquid nitrogen.
$1,295.78
Specifications
Description ThermalTray Thermo-conductive platform
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Type Price Quantity & Availability  
07210075
View Documents Promotion Details
Corning™
432075
Water bath HP, High Profile
Case for $1,295.78
Only null left
Description
Description
Corning ThermalTray™ thermo-conductive platforms support CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
Specifications
Specifications
ThermalTray Thermo-conductive platform