Learn More
Research Products Corporation ThermalSeal RT Films for Real-Time PCR, 2ml thick, Non-Sterile, 100 per Package
Catalog No. 50550328
Encompass
ThermalSeal RT Films are optically transparent, non-absorbing, non-fluorescing and designed for use in Real-Time PCR applications. Films feature a smooth adhesive allowing extreme optical clarity and superior light transmission. End tabs and split backing allow accurate plate positioning. Available 100 per package.  ThermalSeal RT is a Trademark of Excel Scientific, Inc.
Ideal for optical assay
Brilliant ultra-high optical clarity
RNase, DNase and nucleic acid free
Withstand -40° to +120 °C
Catalog No. 50550328 Supplier Research Products Corporation Supplier No. RPI 252000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ThermalSeal RT Films are optically transparent, non-absorbing, non-fluorescing and designed for use in Real-Time PCR applications. Films feature a smooth adhesive allowing extreme optical clarity and superior light transmission. End tabs and split backing allow accurate plate positioning. Available 100 per package.  ThermalSeal RT is a Trademark of Excel Scientific, Inc.
Ideal for optical assay
Brilliant ultra-high optical clarity
RNase, DNase and nucleic acid free
Withstand -40° to +120 °C