missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Thermal Transfer Well Plate Labels

Thermal Transfer Well Plate Labels are ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.

$102.01 - $219.60

Specifications

Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Color White
Display Consignment
Format Label
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Height (English) Height (Metric) Material Price Quantity  
Catalog Number Mfr. No. Quantity Height (English) Height (Metric) Material Price Quantity  
11-877-94
View Documents
Brady™
62374
Each (2500 labels/roll) 0.2 in. 0.5 cm Polyester
Each for $102.01
Only null left
 
11-877-97
View Documents
Brady™
THT1514993
Each (3000 labels/roll) 2 in. 50.8 mm Nylon Cloth
Each for $219.60
Only null left
 
Description

Description

  • Printable labels for thermal transfer printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Glossy white finish with permanent acrylic (B-423) meets requirements of a halogen-free material per DIN VDE 0472 part 815 with service temperature of -94 to 230° F (-70 to 110° C)
Specifications

Specifications

Thermal Transfer Printable Labels
White
Label
Thermal Transfer
Non-sterile
1
2.5 in.
Acrylic, Permanent
Consignment
Single-side Printable
Thermal Transfer Printer Label
BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Rectangle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'