missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Thermal Transfer Film
Description
- Thermal transfer printer ribbon
- Available for Interscience thermal printers or DataLink Pro
Specifications
Specifications
| Product Type | Thermal Transfer Printer Ribbon |
| Color | Black |
| For Use With (Equipment) | DataLink Pro |
| Print Technology | Thermal Transfer |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Format | Roll |