missing translation for 'onlineSavingsMsg'
Learn More

Interscience Thermal Printer Paper Rolls

Catalog No. 16948983
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
16948983 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16948983 Supplier Interscience Supplier No. 521110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermal paper rolls are listed as printer accessories for DiluFlow Elite gravimetric dilutors

  • Thermal paper rolls are listed among the DiluFlow Elite accessories.
  • The rolls are intended for the equipment thermal printer.
  • The accessory supports printed documentation in laboratory workflows.

Specifications

Autoclavable Not Autoclavable
Adhesive Type Non-adhesive
Coating None
Color White
Display Blank
Finish Matte
Format Roll
Material Paper
Print Technology Direct Thermal
Product Type Thermal Paper
Sterility Non-sterile
For Use With (Equipment) Thermal Printers
Languages No Text
No. per Pack 2 Rolls
Show More Show Less