The Thermal Printer DPU-S445 is a thermal line dot printer with high printing speed while producing a clear type face. This allows the Thermal Printer DPU-S445 to print out scanned images. As an alternative to using USB sticks, the automatic print-out function of the DPU-S445 printer can be used to quickly obtain the results. Automatic print-out of results Easy and direct connection via USB Compact and light-weight (approx. 0.5 kg) Easy paper operation Max. printing speed: 90 mm/sec. Number of dots/line: 832 Resolution (dots/mm): 8