Learn More
Applied Biosystems™ Thermal Cycler Fleet Control Software
Catalog No. A40070
Encompass
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
A40070 1 each
1 options
Catalog No. A40070 Supplier Applied Biosystems™ Supplier No. A40070
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stay in control with Thermal Cycler Fleet Control Software.
Applied Biosystems™ Fleet Control Software is as intuitive and easy to use as our thermal cyclers. With this tool, you’ll be able to view and control all Applied Biosystems™ ProFlex™ PCR Systems, Automated Thermal Cyclers, and SimpliAmp™ Thermal Cyclers in your facility.
Secure—Fleet Control Software comes installed on a server that sits securely behind your firewall
Flexible—create custom permissions and rules for users, methods, and instruments
Powerful—quickly view your thermal cycler inventory and availability right from the dashboard
Convenient—control your thermal cyclers, reports, methods, and firmware upgrades from your office, another lab computer, or any other networked device
Specifications
Voltage 100/240 V
Product Line Applied Biosystems
Display Type 19.5 in. Wide-Viewing Angle
Software Type Instrument Mangement and Control
Product Type Thermal Cycler Fleet Control Software
For Use With (Application) PCR
Quantity 1 each
How do I set up methods on my Thermal Cycler Fleet Control Software?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on setting up runs and methods.
How do I connect my Thermal Cycler Fleet Control Software to the network?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on network configuration.
How do I create and manage users in the Thermal Cycler Fleet Control Software?
Within the Thermal Cycler Fleet Control Software, go to Settings, then Help Tutorials, where you can find tutorials on creating users and assigning user permissions.
Where is the manual for the Thermal Cycler Fleet Control Software?
The Quick Resource Card for the Fleet Control Software can be found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100077923_ThermalCyclerFleetSftwre_QR.pdf.
How can I monitor my thermal cycler remotely without being on the Cloud?
The Thermal Cycler Fleet Control Software allows monitoring of the following instruments from within a firewall:
SimpliAmp Thermal Cycler
ProFlex PCR System
Veriti Thermal Cycler
VeritiPro Thermal Cycler
Automated Thermal Cycler (ATC)
Which thermocyclers can be controlled using the Thermal Cycler Fleet Control Software?
The Thermal Cycler Fleet Control Software is compatible with the following instruments:
- SimpliAmp Thermal Cycler
- ProFlex PCR System
- Veriti Thermal Cycler
- VeritiPro Thermal Cycler
- Automated Thermal Cycler (ATC)
- MiniAmp Thermal Cycler
- MiniAmp Plus Thermal Cycler
For Research Use Only. Not for use in diagnostic procedures.